Learn More
Aviva Systems Biology Corp GPX3 Antibody - N-terminal region (ARP41491_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41491_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GPX3. Alternate names: GPx-P, GSHPx-3, GSHPx-PApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-57881