Learn More
AdipoGen GPX3 human rec
SDP
Catalog No. 502563048
Encompass_Preferred
Protein. Human GPX3 aa 21-226 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS, pH7.2. Purity >90% SDS-PAGE. Glutathione peroxidase 3 GPX3 is a selenium-dependent extracellular enzyme, protecting cells and enzymes from oxidative damage by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxides. GPX3 is involved in inflammatory bowel disease, in asthma and in diabetes. The activity of GPX3 is significantly reduced in the plasma and tissue of cancer patients. Recently, GPX3 was suggested to serve as a molecular marker for the diagnosis of clear cell type ovarian adenocarcinoma.
Catalog No. 50-256-3048 Supplier AdipoGen Supplier No. AG40A0037C050
Protein. Human GPX3 aa 21-226 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS, pH7.2. Purity >90% SDS-PAGE. Glutathione peroxidase 3 GPX3 is a selenium-dependent extracellular enzyme, protecting cells and enzymes from oxidative damage by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxides. GPX3 is involved in inflammatory bowel disease, in asthma and in diabetes. The activity of GPX3 is significantly reduced in the plasma and tissue of cancer patients. Recently, GPX3 was suggested to serve as a molecular marker for the diagnosis of clear cell type ovarian adenocarcinoma.