Learn More
X3 Industrial Grade Vinyl Gloves

Description
- Constructed from lightweight, 3 mil vinyl
- Tactile sensitivity, comfort, and dexterity at a lower price than nitrile or latex
- Vinyl is a synthetic material made from polyvinyl chloride with a looser fit and smooth, comfortable feel
- Dependable barrier protection against common cleaning solutions, hair dyes, and other non-hazardous materials
- Suited for work environments such as food service, painting, janitorial, sanitation, plumbing, salon & beauty
- Ideal for light-industrial applications where frequent glove changes are required
- Latex-free for those allergic to natural rubber
- Powder free to prevent messy residue
Specifications
Specifications
| Applications | Industrial |
| AQL | 4 |
| Color | Clear |
| Disposable | Single-use |
| Exterior Finish | Smooth |
| Finger Thickness (English) | 3 mil |
| Finger Thickness (Metric) | 0.08 mm |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Vinyl |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.