Learn More
AdipoGen GPX3 pAb
SDP
Supplier:  AdipoGen AG25A0073C100
Encompass_Preferred
Polyclonal Antibody. Recognizes mouse GPX3. Detects a band of ~20-27kDa by Western blot. Weakly cross-reacts with human GPX3. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Glutathione peroxidase 3 GPX3 GSHPX3 is a selenium-dependent extracellular enzyme, protecting cells and enzymes from oxidative damage by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxides. GPX3 is involved in inflammatory bowel disease, in asthma and in diabetes. The activity of GPX3 is significantly reduced in the plasma and tissue of cancer patients. Recently, GPX3 was suggested to serve as a molecular marker for the diagnosis of clear cell type ovarian adenocarcinoma.
Catalog No. 50-256-3242