Learn More
Cayman Chemical GPX4 asy Buffer10x 1ea
SDP
Catalog No. 502896629
Encompass_Preferred
GPX4 Assay Buffer (10x) has been tested and formulated to work exclusively with Cayman’s GPX4 Inhibitor Screening Assay Kit (Item No. 701880).
Catalog No. 50-289-6629 Supplier Cayman Chemical Supplier No. 7018811 EA
GPX4 Assay Buffer (10x) has been tested and formulated to work exclusively with Cayman’s GPX4 Inhibitor Screening Assay Kit (Item No. 701880).