Learn More
Cayman Chemical GPX4 Glutathione Reductase, 1 ea
SDP
Catalog No. 502896165
Encompass_Preferred
GPX4 Glutathione Reductase has been tested and formulated to work exclusively with Cayman’s GPX4 Inhibitor Screening Assay Kit (Item No. 701880).
Catalog No. 50-289-6165 Supplier Cayman Chemical Supplier No. 7018841 EA
GPX4 Glutathione Reductase has been tested and formulated to work exclusively with Cayman’s GPX4 Inhibitor Screening Assay Kit (Item No. 701880).