Learn More
Medchemexpress LLC GPX4-IN-3 | 2761004-85-5 | 99.8% | C29H24ClN3O3S | 100 MG
SDP
Catalog No. 5000354530
Encompass_Preferred
GPX4-IN-3 is a potent and selective glutathione peroxidase 4 (GPX4) inhibitor and a selective ferroptosis inducer. It demonstrates significant inhibition of GPX4 and effectively induces ferroptosis in various cell lines, showing a strong ability to increase lipid peroxides. This compound also exhibits antitumor activity and good biological safety in preclinical models, effectively suppressing tumor growth.
  • Potent glutathione peroxidase 4 (GPX4) inhibitor
  • Selective ferroptosis inducer
  • Increases lipid peroxide levels
  • Induces ferroptosis with satisfactory selectivity
  • Increases reactive oxygen species (ROS) levels in 4T1 cells
  • Exhibits antitumor activity
  • Good biological safety in vivo
  • Suppresses tumor growth in mouse xenograft models
Catalog No. 50-003-54530 Supplier Medchemexpress LLC Supplier No. HY141809100MG
GPX4-IN-3 is a potent and selective glutathione peroxidase 4 (GPX4) inhibitor and a selective ferroptosis inducer. It demonstrates significant inhibition of GPX4 and effectively induces ferroptosis in various cell lines, showing a strong ability to increase lipid peroxides. This compound also exhibits antitumor activity and good biological safety in preclinical models, effectively suppressing tumor growth.
  • Potent glutathione peroxidase 4 (GPX4) inhibitor
  • Selective ferroptosis inducer
  • Increases lipid peroxide levels
  • Induces ferroptosis with satisfactory selectivity
  • Increases reactive oxygen species (ROS) levels in 4T1 cells
  • Exhibits antitumor activity
  • Good biological safety in vivo
  • Suppresses tumor growth in mouse xenograft models