Learn More
Medchemexpress LLC HY-141809 5mg , GPX4-IN-3 CAS:2761004-85-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1418095MG
Encompass_Preferred
HY-141809 5mg GPX4-IN-3 CAS: 2761004-85-5 GPX4-IN-3 (26a) is a potent glutathione peroxidase 4 (GPX4) inhibitor as a selective ferroptosis inducer. GPX4-IN-3 (26a) exhibits 71.7% inhibition for GPX4 with 1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1457