Learn More
Cayman Chemical GPX4 InhIbItr Screeng as 96Wel
SDP
Catalog No. 502892728
Encompass_Preferred
Cayman's GPX4 Inhibitor Screening Assay Kit provides a robust and EAsy-to-use platform for identifying novel inhibitors of human GPX4, a negative regulator of the ferroptosis pathway. The assay measures GPX4 indirectly by a coupled reaction with glutathione reductase (GR). Oxidized glutathione (GSSG), produced upon reduction of hydroperoxide by GPX4, is recycled to its reduced state by GR and NADPH. The oxidation of NADPH to NADP+ is accompanied by a decrease in absorbance at 340 nm. The rate of decrease in A340 is directly proportional to GPX4 activity. The GPX4 inhibitor ML-162 is included as a positive control
Catalog No. 50-289-2728 Supplier Cayman Chemical Supplier No. 70188096 WELL
Add to Cart
Add to Cart
Cayman's GPX4 Inhibitor Screening Assay Kit provides a robust and EAsy-to-use platform for identifying novel inhibitors of human GPX4, a negative regulator of the ferroptosis pathway. The assay measures GPX4 indirectly by a coupled reaction with glutathione reductase (GR). Oxidized glutathione (GSSG), produced upon reduction of hydroperoxide by GPX4, is recycled to its reduced state by GR and NADPH. The oxidation of NADPH to NADP+ is accompanied by a decrease in absorbance at 340 nm. The rate of decrease in A340 is directly proportional to GPX4 activity. The GPX4 inhibitor ML-162 is included as a positive control