Learn More
Enzo Life Sciences Ganglioside GQ1b. tetrasodium salt (bovine brain) (0.5mg). CAS: 68652-37-9
SDP
Supplier:  Enzo Life Sciences ALX302012MC05
Encompass_Preferred
Specifically promotes neural differentiation of human neuroblastoma cells. Alternative name: GQ1b . 4Na (bovine brain), Tetrasialoganglioside GQ1b . 4 Na (bovine brain). Purity: ≥98% (TLC). Appearance: Lyophilized. Solubility: Soluble in water, biological buffers or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8653