Learn More
Medchemexpress LLC Gr 127935 hydrochloride | 148642-42-6 | MFCD08056244 | 99.4% | 534.05 | C29H32ClN5O3 | 10 MG
SDP
Catalog No. 5000212309
Encompass_Preferred
GR127935 hydrochloride is a potent, orally active antagonist of 5-HT1D and 5-HT1B serotonin receptors used in neurological and pharmacological research. Supplied as a white to off-white solid, it is characterized by high purity and documented solubility and storage recommendations for reliable experimental use.
  • High purity suitable for research applications.
  • Potent 5-HT1D and 5-HT1B receptor antagonism for receptor pharmacology studies.
  • Supplied in small milligram quantities for in vitro and in vivo dosing.
  • Soluble in DMSO; follow recommended handling and storage.
  • Documentation includes datasheet, certificate of analysis, and safety data sheet for traceability.
Catalog No. 50-002-12309 Supplier Medchemexpress LLC Supplier No. HY10079410MG
GR127935 hydrochloride is a potent, orally active antagonist of 5-HT1D and 5-HT1B serotonin receptors used in neurological and pharmacological research. Supplied as a white to off-white solid, it is characterized by high purity and documented solubility and storage recommendations for reliable experimental use.
  • High purity suitable for research applications.
  • Potent 5-HT1D and 5-HT1B receptor antagonism for receptor pharmacology studies.
  • Supplied in small milligram quantities for in vitro and in vivo dosing.
  • Soluble in DMSO; follow recommended handling and storage.
  • Documentation includes datasheet, certificate of analysis, and safety data sheet for traceability.