Learn More
Abcam GR 46611, 5-HT1B/1D agonist, 50MG
Catalog No. 502707459
Encompass_Preferred
MW 377.49 g/mol, Purity >97%. 5-HT₁B/₁D agonist. 10-fold higher selectivity over 5-HT₁A. Displays centrally mediated effects upon systemic administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7459 Supplier Abcam Supplier No. AB12058650MG
MW 377.49 g/mol, Purity >97%. 5-HT₁B/₁D agonist. 10-fold higher selectivity over 5-HT₁A. Displays centrally mediated effects upon systemic administration.
The product is subject to the following: Abcam Restricted Use Statement