Learn More
Medchemexpress LLC GR 55562 dihydrochloride | 159533-25-2 | 99.0% | 448.39 | C23H27Cl2N3O2 | 10 MG
SDP
Catalog No. 5000212838
Encompass_Preferred
GR 55562 dihydrochloride is a research-grade small molecule that acts as a selective 5-HT1B (serotonin) receptor antagonist for preclinical and in vitro studies probing 5-HT1B-mediated signaling and neurological disease mechanisms.
  • Selective 5-HT1B receptor antagonist.
  • High purity (99.0%).
  • Solid form suitable for analytical and biological assays.
  • Store under nitrogen, away from moisture; solvent storage -80°C (6 months) or -20°C (1 month).
  • Available in 5 mg and 10 mg pack sizes.
Catalog No. 50-002-12838 Supplier Medchemexpress LLC Supplier No. HY101367A10MG
GR 55562 dihydrochloride is a research-grade small molecule that acts as a selective 5-HT1B (serotonin) receptor antagonist for preclinical and in vitro studies probing 5-HT1B-mediated signaling and neurological disease mechanisms.
  • Selective 5-HT1B receptor antagonist.
  • High purity (99.0%).
  • Solid form suitable for analytical and biological assays.
  • Store under nitrogen, away from moisture; solvent storage -80°C (6 months) or -20°C (1 month).
  • Available in 5 mg and 10 mg pack sizes.