Learn More
Medchemexpress LLC Gr 55562 dihydrochloride | 159533-25-2 | 99.0% | 448.39 | C23H27Cl2N3O2 | 5 MG
SDP
Catalog No. 5000213207
Encompass_Preferred
GR 55562 dihydrochloride is a selective serotonin 5-HT1B and 5-HT1D receptor antagonist supplied as the dihydrochloride salt for research use. It is commonly used in receptor pharmacology and behavioral studies to probe serotonergic mechanisms in vitro and in vivo.
  • Selective 5-HT1B/5-HT1D receptor antagonist.
  • Suitable for in vivo and in vitro studies.
  • High purity (99.0%).
  • Provided as the dihydrochloride salt for improved solubility.
  • Available in small quantities for research use (5 MG, 10 MG).
  • Includes documentation: datasheet, COA, and SDS.
Catalog No. 50-002-13207 Supplier Medchemexpress LLC Supplier No. HY101367A5MG
GR 55562 dihydrochloride is a selective serotonin 5-HT1B and 5-HT1D receptor antagonist supplied as the dihydrochloride salt for research use. It is commonly used in receptor pharmacology and behavioral studies to probe serotonergic mechanisms in vitro and in vivo.
  • Selective 5-HT1B/5-HT1D receptor antagonist.
  • Suitable for in vivo and in vitro studies.
  • High purity (99.0%).
  • Provided as the dihydrochloride salt for improved solubility.
  • Available in small quantities for research use (5 MG, 10 MG).
  • Includes documentation: datasheet, COA, and SDS.