Learn More
MSC Value Hex Head Screw, 1/2-13, 9 in, Grade 8, 10/Pk.
Catalog No. 502866088
Encompass
This hex head cap screw is engineered for demanding applications, providing secure fastening with high tensile strength and corrosion resistance. It is suitable for a wide range of industrial and construction uses.
  • Manufactured from durable steel
  • Grade 8 material for high strength
  • Yellow zinc dichromate finish for corrosion protection
  • Features 1/2-13 thread size and a 9 in length under head
  • Partially threaded for application versatility
  • Conforms to SAE J429 and ASME B18.2.1 standards
Catalog No. 50-286-6088 Supplier MSC Supplier No. 70985536
This hex head cap screw is engineered for demanding applications, providing secure fastening with high tensile strength and corrosion resistance. It is suitable for a wide range of industrial and construction uses.
  • Manufactured from durable steel
  • Grade 8 material for high strength
  • Yellow zinc dichromate finish for corrosion protection
  • Features 1/2-13 thread size and a 9 in length under head
  • Partially threaded for application versatility
  • Conforms to SAE J429 and ASME B18.2.1 standards