Learn More
MSC Value Collection Cap Screw, 1/4-20, 1 in, 50 Pack
Catalog No. 502870358
Encompass
Hex head cap screws are designed for heavy-duty fastening applications, offering high strength and durability. This grade 8 steel screw features a yellow zinc dichromate finish for corrosion resistance and is fully threaded for secure engagement.
  • High-strength grade 8 steel construction
  • Yellow zinc dichromate finish provides corrosion resistance
  • Fully threaded for maximum engagement
  • Complies with SAE J429 and ASME B18.2.1 standards
  • Hex head design for easy installation with a 7/16 in hex key
  • Ideal for demanding fastening requirements
Catalog No. 50-287-0358 Supplier MSC Supplier No. 80930050
Hex head cap screws are designed for heavy-duty fastening applications, offering high strength and durability. This grade 8 steel screw features a yellow zinc dichromate finish for corrosion resistance and is fully threaded for secure engagement.
  • High-strength grade 8 steel construction
  • Yellow zinc dichromate finish provides corrosion resistance
  • Fully threaded for maximum engagement
  • Complies with SAE J429 and ASME B18.2.1 standards
  • Hex head design for easy installation with a 7/16 in hex key
  • Ideal for demanding fastening requirements