Learn More
MSC Value Collection Hex Head Cap Screw, 1/4-20, Grade 8, 50/Pk.
Catalog No. 502870360
Encompass
This hex head cap screw is engineered for demanding applications, featuring a robust Grade 8 steel construction for superior strength. Its yellow zinc dichromate finish provides enhanced corrosion resistance, making it suitable for environments where durability and protection are paramount.
  • Designed to meet ASME B18.2.1 standards
  • Fully threaded for secure fastening
  • Standard hex head for easy installation and removal
  • Offers high tensile strength for heavy-duty use
Catalog No. 50-287-0360 Supplier MSC Supplier No. 80930084
This hex head cap screw is engineered for demanding applications, featuring a robust Grade 8 steel construction for superior strength. Its yellow zinc dichromate finish provides enhanced corrosion resistance, making it suitable for environments where durability and protection are paramount.
  • Designed to meet ASME B18.2.1 standards
  • Fully threaded for secure fastening
  • Standard hex head for easy installation and removal
  • Offers high tensile strength for heavy-duty use