Learn More
MSC Value Collection Hex Cap Screw, 3/4-10 Grade 8, 25 Pack
Catalog No. 502867519
Encompass
Hex head cap screws are designed for heavy-duty fastening applications, offering secure and reliable connections. This specific screw features a 3/4-10 thread size and 1 in length under head, constructed from Grade 8 steel for high strength and durability.
  • Fully threaded design for maximum engagement
  • Features 1-1/8 in hex for secure tightening
  • Bright/uncoated finish suitable for various environments
  • Manufactured to ASME B18.2.1 standards for quality assurance
Catalog No. 50-286-7519 Supplier MSC Supplier No. 70985528
Hex head cap screws are designed for heavy-duty fastening applications, offering secure and reliable connections. This specific screw features a 3/4-10 thread size and 1 in length under head, constructed from Grade 8 steel for high strength and durability.
  • Fully threaded design for maximum engagement
  • Features 1-1/8 in hex for secure tightening
  • Bright/uncoated finish suitable for various environments
  • Manufactured to ASME B18.2.1 standards for quality assurance