Learn More
CPC Scientific Boc-Arg-Ala-DTrp-Phe-DPro-Pro-Nle-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109886
Encompass
Highly potent, selective long acting, and in vivo NK2 receptor antagonist.SEQUENCE: Boc-Arg-Ala-DTrp-Phe-DPro-Pro-Nle-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Boc-RA-DTrp-F-DPro-P-Nle-NH2MOLECULAR FORMULA: C50H72N12O9MOLECULAR WEIGHT: 985.2STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: NeuropeptidesREFERENCES:A.B. McElroy et al., J. Med. Chem., 35, 2582 (1992)SKU(s): NEUP-004A, NEUP-004B
Catalog No. 50-210-9886 Supplier CPC Scientific Supplier No. NEUP004A
Highly potent, selective long acting, and in vivo NK2 receptor antagonist.SEQUENCE: Boc-Arg-Ala-DTrp-Phe-DPro-Pro-Nle-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Boc-RA-DTrp-F-DPro-P-Nle-NH2MOLECULAR FORMULA: C50H72N12O9MOLECULAR WEIGHT: 985.2STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: NeuropeptidesREFERENCES:A.B. McElroy et al., J. Med. Chem., 35, 2582 (1992)SKU(s): NEUP-004A, NEUP-004B