Learn More
CPC Scientific Boc-Ala-Ala-DTrp-Phe-DPro-Pro-Nle-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP005B
Encompass
Highly potent and selective NK2 receptor antagonist.SEQUENCE: Boc-Ala-Ala-DTrp-Phe-DPro-Pro-Nle-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Boc-AA-DTrp-F-DPro-P-Nle-NH2MOLECULAR FORMULA: C47H65N9O9MOLECULAR WEIGHT: 900.1STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: NeuropeptidesREFERENCES:A.B. McElroy et al., J. Med. Chem., 35 2582 (1992)SKU(s): NEUP-005A, NEUP-005B
Catalog No. 50-210-9889