Learn More
Apexbio Technology LLC GR 89696 fumarate 126766-32-3 10mg
SDP
Catalog No. 5000089714
Encompass_Preferred
GR 89696 fumarate (CAS 126766-32-3) is a selective agonist of the 2-opioid receptor By specifically activating 2 subtypes this small molecule modulates neural signaling pathways implicated in sensory processing particularly pain and pruritus Experimental studies have demonstrated that GR 89696 fumarate induces physiological effects in the nervous system through 2-opioid receptor activation This compound has been widely utilized as a pharmacological tool in the investigation of neurocutaneous disorders and the underlying mechanisms of opioid receptor-mediated modulation of somatosensory functions
Catalog No. 50-000-89714 Supplier Apexbio Technology LLC Supplier No. B675710
GR 89696 fumarate (CAS 126766-32-3) is a selective agonist of the 2-opioid receptor By specifically activating 2 subtypes this small molecule modulates neural signaling pathways implicated in sensory processing particularly pain and pruritus Experimental studies have demonstrated that GR 89696 fumarate induces physiological effects in the nervous system through 2-opioid receptor activation This compound has been widely utilized as a pharmacological tool in the investigation of neurocutaneous disorders and the underlying mechanisms of opioid receptor-mediated modulation of somatosensory functions