Learn More
CPC Scientific PhCO-Ala-Ala-DTrp-Phe-DPro-Pro-Nle-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific NEUP006A
Encompass
Selective NK-2 tachykinin receptor antagonist.SEQUENCE: PhCO-Ala-Ala-DTrp-Phe-DPro-Pro-Nle-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: PhCO-AA-DTrp-F-DPro-P-Nle-NH2MOLECULAR FORMULA: C49H61N9O8MOLECULAR WEIGHT: 904.06STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [141636-65-9]SYNONYMS: RESEARCH AREA: NeuropeptidesREFERENCES:A.B. McElroy et al., J. Med. Chem., 35, 2582 (1992)SKU(s): NEUP-006A, NEUP-006B
Catalog No. 50-210-9890