Learn More
Apexbio Technology LLC GR 94800 TFA 5mg
SDP
Catalog No. 5000064228
Encompass_Preferred
GR 94800 TFA is a selective antagonist targeting neuropeptide Y (NPY) Y2 receptors Neuropeptide Y signaling via Y2 receptor subtypes participates in energy balance appetite modulation mood regulation anxiety-related behaviors and neuronal response to stress GR 94800 TFA specifically inhibits NPY Y2 receptor-mediated signaling thereby facilitating studies on the physiological and behavioral outcomes regulated by this receptor pathway It is commonly utilized by biomedical researchers investigating functions of NPY systems in feeding control emotional responses anxiety and depressive behaviors as well as cognition-related processes such as learning and memory Additionally this compound serves as an investigative tool to elucidate downstream signaling pathways and potential pharmacological targets linked to the Y2 receptor
Catalog No. 50-000-64228 Supplier Apexbio Technology LLC Supplier No. B87875
GR 94800 TFA is a selective antagonist targeting neuropeptide Y (NPY) Y2 receptors Neuropeptide Y signaling via Y2 receptor subtypes participates in energy balance appetite modulation mood regulation anxiety-related behaviors and neuronal response to stress GR 94800 TFA specifically inhibits NPY Y2 receptor-mediated signaling thereby facilitating studies on the physiological and behavioral outcomes regulated by this receptor pathway It is commonly utilized by biomedical researchers investigating functions of NPY systems in feeding control emotional responses anxiety and depressive behaviors as well as cognition-related processes such as learning and memory Additionally this compound serves as an investigative tool to elucidate downstream signaling pathways and potential pharmacological targets linked to the Y2 receptor