Learn More
TARGETMOL CHEMICALS INC GR-95168 1MG
SDP
Catalog No. 502879893
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. GR-95168 ((+)-C-BVDU) is an antiviral compound for the study of herpes simplex virus infections type 1 and type 2. Purity 99.73%
Catalog No. 50-287-9893 Supplier TARGETMOL CHEMICALS INC Supplier No. T678001MG
Add to Cart
Add to Cart
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. GR-95168 ((+)-C-BVDU) is an antiviral compound for the study of herpes simplex virus infections type 1 and type 2. Purity 99.73%