Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GR ACS Acetic Anhydride, MilliporeSigma™

Catalog No. MAX00805 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
3.78 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX00805 3.78 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX00805 Supplier MilliporeSigma Supplier No. AX00805
Add to Cart
Add to Cart

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Melting Point -72.8°C
Boiling Point 140°C
Flash Point 48.85°C (119.9°F)
Packaging Glass Bottle
Quantity 3.78 L
UN Number UN 1715
Solubility Information Water: Partially soluble
Formula Weight 102.09
Percent Purity ≥99.0%
Grade ACS
Physical Form Liquid
Chemical Name or Material Acetic Anhydride
Show More Show Less
DOTInformation DOT Class: 3 (8), Packaging Group: II
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.