Learn More
Enzo Life Sciences Glucocorticoid receptor alpha polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ALX210277C050
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 755-771 (C755EIITNQIPKYSN GNIKK771) of human GRα (glucocorticoid receptor α). Species reactivity: Human, Mouse, Rat. Applications: ICC, IP, WB ChIP. Purity: Epitope-affinity purified IgG. Formulation: Lyophilized. Contains 0.05% sodium azide.
Catalog No. 50-200-8101