Learn More
Braintree Scientific GRAEFE Fixation Forceps
SDP
Catalog No. 501954837
Encompass
GRAEFE Fixation Forceps 4-3/8" (11.2 cm), standard jaws 4.5 mm wide, with fine teeth **Sold for research purposes only and not for human use**
Catalog No. 50-195-4837 Supplier Braintree Scientific Supplier No. FC850
GRAEFE Fixation Forceps 4-3/8" (11.2 cm), standard jaws 4.5 mm wide, with fine teeth **Sold for research purposes only and not for human use**