Learn More
Foxx Life Sciences Borosil Reusable Class A Mohr (Measuring) Pipettes (Pipets) with Permanent Amber Graduations and Individual Serial Numbers, Batch Certificate, 25mL (0.20mL Intervals), 10/CS
Catalog No. 501542883
Encompass_Preferred
Borosil Reusable Class A Mohr (Measuring) Pipettes are graduated using a permanent amber treatment that will not blur, wear, or wash off even after long-term use. Each pipette is individually calibrated by automatic, laser-controlled calibration equipment. Manufactured with consistent wall thickness. Reusable. Uniform inner bore diameter for accuracy between different readings. Beveled tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil Mohr Pipettes are graduated for delivery from the zero mark to the final graduation mark. Borosil Class A Mohr Pipettes comply with EN ISO 835. Avail. from 0.1mL to 25mL. 1mL, 2mL, and 5mL sizes Avail. in two different graduation intervals. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices.
Catalog No. 50-154-2883 Supplier Foxx Life Sciences Supplier No. 7059P09A
Borosil Reusable Class A Mohr (Measuring) Pipettes are graduated using a permanent amber treatment that will not blur, wear, or wash off even after long-term use. Each pipette is individually calibrated by automatic, laser-controlled calibration equipment. Manufactured with consistent wall thickness. Reusable. Uniform inner bore diameter for accuracy between different readings. Beveled tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil Mohr Pipettes are graduated for delivery from the zero mark to the final graduation mark. Borosil Class A Mohr Pipettes comply with EN ISO 835. Avail. from 0.1mL to 25mL. 1mL, 2mL, and 5mL sizes Avail. in two different graduation intervals. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices.