Learn More
Cayman Chemical GR127935 (hydrochloride), Purity: ≥98%, CAS: 148642-42-6, 50 mg
SDP
Catalog No. 502988933
Encompass_Preferred
A 5-HT1B and 5-HT1D receptor antagonist (IC50s = 3.16, 1.26, and 0.13 nM, respectively); selective for the 5-HT1B and 5-HT1D receptors over other 5-HT receptors (IC50s = 39.81->10,000 nM in radioligand binding assays); enhances memory consolidation in an autoshaping learning task in rats when administered post-training at 10 mg/kg; has been used to elucidate the 5-HT receptor subtypes involved in the anti-inflammatory activity of sumatriptan
Catalog No. 50-298-8933 Supplier Cayman Chemical Supplier No. 2965150 MG
A 5-HT1B and 5-HT1D receptor antagonist (IC50s = 3.16, 1.26, and 0.13 nM, respectively); selective for the 5-HT1B and 5-HT1D receptors over other 5-HT receptors (IC50s = 39.81->10,000 nM in radioligand binding assays); enhances memory consolidation in an autoshaping learning task in rats when administered post-training at 10 mg/kg; has been used to elucidate the 5-HT receptor subtypes involved in the anti-inflammatory activity of sumatriptan