Learn More
Medchemexpress LLC HY-110390 5mg Medchemexpress, GR148672X CAS:263890-70-6 Purity:>98%
SDP
Catalog No. 501968796
Encompass_Preferred
Medchemexpress, HY-110390 5mg GR148672X CAS:263890-70-6 GR148672X is a triacyglycerol hydrolase (TGH) inhibitor with an IC50 of 4 nM extracted from patent WO 2001016358 A2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8796 Supplier Medchemexpress LLC Supplier No. HY1103905MG
Medchemexpress, HY-110390 5mg GR148672X CAS:263890-70-6 GR148672X is a triacyglycerol hydrolase (TGH) inhibitor with an IC50 of 4 nM extracted from patent WO 2001016358 A2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.