Learn More
Cayman Chemical GR89696fumarate 10mg
SDP
Catalog No. 502942763
Encompass_Preferred
A κ-opioid receptor agonist; inhibits twitch responses in isolated mouse vas deferens (IC50 = 0.12 nM); reduces acetylcholine-induced writhing in mice (ED50 = 0.0005 mg/kg); reduces hippocampal neuronal loss in a gerbil model of cerebral ischemia induced by bilateral carotid artery occlusion at 10 and 30 mg/kg
Catalog No. 50-294-2763 Supplier Cayman Chemical Supplier No. 4014510 MG
A κ-opioid receptor agonist; inhibits twitch responses in isolated mouse vas deferens (IC50 = 0.12 nM); reduces acetylcholine-induced writhing in mice (ED50 = 0.0005 mg/kg); reduces hippocampal neuronal loss in a gerbil model of cerebral ischemia induced by bilateral carotid artery occlusion at 10 and 30 mg/kg