Learn More
TARGETMOL CHEMICALS INC GRA EX-25 5MG
SDP
Catalog No. 502580275
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GRA Ex-25 (GRA-Ex-25) is an inhibitor of glucagon receptor. purity: 98%
Catalog No. 50-258-0275 Supplier TARGETMOL CHEMICALS INC Supplier No. T34225MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GRA Ex-25 (GRA-Ex-25) is an inhibitor of glucagon receptor. purity: 98%