Learn More
Foxx Life Sciences Borosil® Reusable Class A Mohr (Measuring) Pipettes (Pipets) with Permanent Amber Graduations and Individual Serial Numbers, Individual Certificate, 0.1mL ( 0.01ml Intervals) , 10/CS
Supplier:  Foxx Life Sciences 7059P41D
Encompass_Preferred
Borosil® Reusable Class A Mohr (Measuring) Pipettes are graduated using a permanent amber treatment that will not blur, wear, or wash off even after long-term use. Each pipette is individually calibrated by automatic, laser-controlled calibration equipment. Manufactured with consistent wall thickness. Reusable. Uniform inner bore diameter for accuracy between different readings. Beveled tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil® Mohr Pipettes are graduated for delivery from the zero mark to the final graduation mark. Borosil® Class A Mohr Pipettes comply with EN ISO 835. Available from 0.1mL to 25mL. 1mL, 2mL, and 5mL sizes available in two different graduation intervals. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices.
Catalog No. 50-172-0312