Learn More
High-Tech Conversions Grab-EEZ Stainless Steel Dispenser
Catalog No. 17010306
Change view
Click to view available options
Mounting:
Wall Mount
Catalog No. Mounting
17-010-306 Wall Mount
1 options
Catalog No. 17-010-306 Supplier High-Tech Conversions Supplier No. GEDISPSS
Add to Cart
Add to Cart
Grab-EEZ Stainless Steel Dispenser ideal for dispensing 9 x 9 cleanroom wipes.
  • Stainless Steel Construction ISO Class 5 (Class 100)
  • Cleanroom Compatible
  • Eliminates wasteful wipe usage and cross-contamination
  • Grab-EEZ Wipe Dispenser GE-DISP-SS, Sold individually Grab-EEZ Wipe Refills GE-NT1-99 9x9, 250/Bag 12 Bags/Case
  • Nonwoven Poly-Cellulose Cleanroom Wipes
Specifications
Product Type Cleanroom Wiper Dispenser
Color Silver
Holds Wipes
Material Stainless Steel
Mounting Wall Mount
No. of Compartments 1
Product Style Top Loading
Sterility Non-sterile