Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs Culturewell
Catalog No. 502901627
Encompass
CultureWell ChamberSLIP 16 NON-Removable Chambered Coverglass 16 Well No. 1.5 German borosilicate coverglass. The product consists of cell culture vessels with a 2x8 format with standard 96-well plate volumes and well spacing.ChamberSLIP 16 Coverslip Non-Removable format has fixed chambers that do not separate from the coverglass for applications that do not require mounting fixed cells.
Catalog No. 50-290-1627 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GBL1123598EA
CultureWell ChamberSLIP 16 NON-Removable Chambered Coverglass 16 Well No. 1.5 German borosilicate coverglass. The product consists of cell culture vessels with a 2x8 format with standard 96-well plate volumes and well spacing.ChamberSLIP 16 Coverslip Non-Removable format has fixed chambers that do not separate from the coverglass for applications that do not require mounting fixed cells.