Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs CultureWell
Catalog No. 502904866
Encompass
CS16 - CultureWell(TM) with Removable Chambered Coverglass - ideal for cell culture and fluorescence imaging applicationsThe 2 x 8 16 chambered coverglass is like a chambered slide with the imaging advantages of a removable coverglass substructure. It is designed with standard 96 well plate volumes and well spacing. The non-cytotoxic silicone well gasket forms a leak-proof seal between the polycarbonate upper structure and the coverglass. CultureWell(TM) chambered coverglass is shipped sterile and ready to use.Coverglass removal is facilitated using a very simple device that reliably separates the parts without the need for excessive force or risk of cover glass breakage. Silicone removal leaves no residue. Components are manufactured and assembled with special orientation features for ease of handling and convenient identification or specimen location after well structure removal.The growth surface is No. 1.5 German borosilicate coverglassInert non-cytotoxic silicone permi
Catalog No. 50-290-4866 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GBL1123588EA
CS16 - CultureWell(TM) with Removable Chambered Coverglass - ideal for cell culture and fluorescence imaging applicationsThe 2 x 8 16 chambered coverglass is like a chambered slide with the imaging advantages of a removable coverglass substructure. It is designed with standard 96 well plate volumes and well spacing. The non-cytotoxic silicone well gasket forms a leak-proof seal between the polycarbonate upper structure and the coverglass. CultureWell(TM) chambered coverglass is shipped sterile and ready to use.Coverglass removal is facilitated using a very simple device that reliably separates the parts without the need for excessive force or risk of cover glass breakage. Silicone removal leaves no residue. Components are manufactured and assembled with special orientation features for ease of handling and convenient identification or specimen location after well structure removal.The growth surface is No. 1.5 German borosilicate coverglassInert non-cytotoxic silicone permi