Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs CultureWell
Catalog No. 502904435
Encompass
CultureWell clear silicone sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.
Catalog No. 50-290-4435 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GBL6644755EA
CultureWell clear silicone sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.