Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs flexWellTM
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GBL20491610EA
Encompass
flexWell(TM) incubation chambers provide a convenient way to conduct binding reactions on protein microarrays using nitrocellulose coated ONCYTE(R) film slides. Silicone flexWell forms wells on slides using a clean release adhesive to isolate up to 16 specimens per slide Slides and flexWell fit into a reusable slide tray organizing wells into the footprint of a SMS compliant microtiter plate Gaskets may be sealed for mixing and to prevent evaporation
Catalog No. 50-290-0854