Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio Labs ONCYTER AVI
Catalog No. 502909041
Encompass
ONCYTE Porous nitrocellulose film slides are comprised of a microporous nitrocellulose film cast on a variety of solid surfaces. They offer numerous advantages over functionalized surface chemistriesIncreased available surface which translates into high protein binding capacity.Fluorescence Signal Amplification via the Tyndall effect that increases the signal intensity providing enhanced sensitivity.Broad dynamic range and low detection limit ONCYTE film slides are manufactured to the highest standards and quality tested they provide excellent reliability and reproducible.
Catalog No. 50-290-9041 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GBL30511620EA
ONCYTE Porous nitrocellulose film slides are comprised of a microporous nitrocellulose film cast on a variety of solid surfaces. They offer numerous advantages over functionalized surface chemistriesIncreased available surface which translates into high protein binding capacity.Fluorescence Signal Amplification via the Tyndall effect that increases the signal intensity providing enhanced sensitivity.Broad dynamic range and low detection limit ONCYTE film slides are manufactured to the highest standards and quality tested they provide excellent reliability and reproducible.