Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs Press-To-Se
Catalog No. 502900369
Encompass
Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Isolators are available in red silicone thickness 0.5 to 2.5mm pre-cut or as sheet material with or without secure seal clean adhesive on one or both surfaces. Nonadhesive isolators are autoclavable and reusable.Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell water-clear silicone which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free HybriSlip covers. Recommended use temperature is -62C to 218C.
Catalog No. 50-290-0369 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GBL66520825EA
Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Isolators are available in red silicone thickness 0.5 to 2.5mm pre-cut or as sheet material with or without secure seal clean adhesive on one or both surfaces. Nonadhesive isolators are autoclavable and reusable.Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell water-clear silicone which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free HybriSlip covers. Recommended use temperature is -62C to 218C.