Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs reusable Cu
Catalog No. 502900344
Encompass
Gaskets are ideal for forming wells on glass microscope slides or in polystyrene dishes. Gaskets are non-sterile and may be sterilized by autoclaving UV or alcohol. Ideal for forming wells on glass microscope slides or in polystyrene dishes and to isolate cells using removable hydrophobic barriers on coverglass.
Catalog No. 50-290-0344 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GBL10325010EA
Gaskets are ideal for forming wells on glass microscope slides or in polystyrene dishes. Gaskets are non-sterile and may be sterilized by autoclaving UV or alcohol. Ideal for forming wells on glass microscope slides or in polystyrene dishes and to isolate cells using removable hydrophobic barriers on coverglass.