Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs SecureSealT
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GBL654002100EA
Encompass
Imaging spacers are ultra-thin adhesive spacers which peel-and-stick to coverglass or microscope slides to confine specimens without compression. Layer multiple spacers to custom build chambers to any depth desired.
Catalog No. 50-290-0223