Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs SecureSealT
Catalog No. 502900942
Encompass
Imaging spacers are ultra-thin adhesive spacers which peel-and-stick to coverglass or microscope slides to confine specimens without compression. Layer multiple spacers to custom build chambers to any depth desired.
Catalog No. 50-290-0942 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GBL654008100EA
Imaging spacers are ultra-thin adhesive spacers which peel-and-stick to coverglass or microscope slides to confine specimens without compression. Layer multiple spacers to custom build chambers to any depth desired.