Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs SecureSealT
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GBL62261425EA
Encompass
SecureSeal Hybridization Chambers are thin silicone-gasketed chambers providing optimal surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and microarrays on glass or film coated slides. These adhesive-backed chambers are designed for single-use very-low volume incubations and are ideal for autoradiographic fluorescent or chemiluminescent end-point applications requiring small enclosed fluid volumes. The SecureSeal Adhesive is a double sided tape with a silicone adhesive surface on one side (covered with a clear PET liner) and an acrylic adhesive on the other surface (covered with a paper liner).SecureSeal chambers form removable peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. The chamber design minimizes friction promotes reagent mixing and facilitates uniform hybridization. Sealable access ports in the chamber surface allow for the addition and removal of reactants. SecureSeal adhesive bonds chambers to g
Catalog No. 50-290-4666