Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs silicone is
Catalog No. 502901664
Encompass
Silicone isolator sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. Where additional sealing is required SecureSeal adhesive on one or both surfaces is recommended.Silicone sheet material is also available inACultureWell water-clear silicone which is highly recommended for sensitive cell culture applications.
Catalog No. 50-290-1664 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GBL6645815EA
Silicone isolator sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. Where additional sealing is required SecureSeal adhesive on one or both surfaces is recommended.Silicone sheet material is also available inACultureWell water-clear silicone which is highly recommended for sensitive cell culture applications.