Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio-Labs wound splin
Catalog No. 502901596
Encompass
Grace Bio-Labs has worked with researchers to develop and manufacture wound splint devices that aid in the understanding of wound and tissue repair for animal research and the testing of new therapies for safety and efficacy.Wound splints are constructed of silicone and include suture sites for increased precision in affixing on or within. These devices enable reproducible and reliable experimental conditions for wound healing in animal research.The splints are offered in a variety of shapes and sizes (visible stitch holes / suture sites 5 mm dia.) shipped sterile and ready to use.
Catalog No. 50-290-1596 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GBLRD47668720EA
Grace Bio-Labs has worked with researchers to develop and manufacture wound splint devices that aid in the understanding of wound and tissue repair for animal research and the testing of new therapies for safety and efficacy.Wound splints are constructed of silicone and include suture sites for increased precision in affixing on or within. These devices enable reproducible and reliable experimental conditions for wound healing in animal research.The splints are offered in a variety of shapes and sizes (visible stitch holes / suture sites 5 mm dia.) shipped sterile and ready to use.