Learn More
Braintree Scientific Grace Procedure/Dissection Board w/LED & magnifier
SDP
Catalog No. 501954878
Encompass
Grace Procedure/Dissection Boards - The New Standard for small animal surgery, procedure, & dissection platforms Boards are made of 1" thick high density polyethylene (HDPE) with excellent chemical resistance, easy-to-clean, solid work surface. Grace Procedure/Dissection Board (GPS1)Non-skid feet for added stability. 12" (L) x 8" (W). Grace Procedure/Dissection Board w/ LED, & Magnifier (GPS2)Two Flexible LED lights - provides direct illumination with no shadows and no heat. Flex arm, non-obstructive rimless magnifying glass (main lens: 2.3X, inset lens: 4.0X). (Allows continuous eye contact with object when moving into magnification area). Non-skid feet for added stability. Universal input power supply (100 240 VAC, 50-60 Hz). Size: 12" (L) x 9.5" (W). **Sold for research purposes only and not for human use**
Catalog No. 50-195-4878 Supplier Braintree Scientific Supplier No. GPS2
Grace Procedure/Dissection Boards - The New Standard for small animal surgery, procedure, & dissection platforms Boards are made of 1" thick high density polyethylene (HDPE) with excellent chemical resistance, easy-to-clean, solid work surface. Grace Procedure/Dissection Board (GPS1)Non-skid feet for added stability. 12" (L) x 8" (W). Grace Procedure/Dissection Board w/ LED, & Magnifier (GPS2)Two Flexible LED lights - provides direct illumination with no shadows and no heat. Flex arm, non-obstructive rimless magnifying glass (main lens: 2.3X, inset lens: 4.0X). (Allows continuous eye contact with object when moving into magnification area). Non-skid feet for added stability. Universal input power supply (100 240 VAC, 50-60 Hz). Size: 12" (L) x 9.5" (W). **Sold for research purposes only and not for human use**