Learn More
Medchemexpress LLC Gracillin | 19083-00-2 | 885.04 | 25 MG
SDP
Catalog No. 5000389864
Encompass_Preferred
Gracillin is a steroidal saponin extracted from plant roots with anti-tumor properties. It can induce cancer cell apoptosis and autophagy.
  • Steroidal saponin.
  • Exhibits anti-tumor properties.
  • Induces cancer cell apoptosis and autophagy.
  • Shows broad-spectrum inhibitory effects on cancer cell viability (e.g., A2780, A549, BXPC-3, Bel-7402, Capan-2, HCT-8, HL-60, HeLa, MCF7, PANC-1, SW1990 cells).
  • Disrupts mitochondrial function by inhibiting ATP production, activating AMPK, and inhibiting oxygen consumption rate (OCR) in H460 and A549 cells.
  • Induces ROS generation in H460, H1299, H226B, and A549 cells.
  • Blocks cell cycle in G1 phase and induces apoptosis and autophagy in A549 cells.
  • Inhibits tumor growth of xenograft tumors in lung (H1299), prostate (DU145), and colorectum (HCT116) cancer cells, as well as patient-derived xenograft colon and lung tumors.
Catalog No. 50-003-89864 Supplier Medchemexpress LLC Supplier No. 25MGHYN0706
Gracillin is a steroidal saponin extracted from plant roots with anti-tumor properties. It can induce cancer cell apoptosis and autophagy.
  • Steroidal saponin.
  • Exhibits anti-tumor properties.
  • Induces cancer cell apoptosis and autophagy.
  • Shows broad-spectrum inhibitory effects on cancer cell viability (e.g., A2780, A549, BXPC-3, Bel-7402, Capan-2, HCT-8, HL-60, HeLa, MCF7, PANC-1, SW1990 cells).
  • Disrupts mitochondrial function by inhibiting ATP production, activating AMPK, and inhibiting oxygen consumption rate (OCR) in H460 and A549 cells.
  • Induces ROS generation in H460, H1299, H226B, and A549 cells.
  • Blocks cell cycle in G1 phase and induces apoptosis and autophagy in A549 cells.
  • Inhibits tumor growth of xenograft tumors in lung (H1299), prostate (DU145), and colorectum (HCT116) cancer cells, as well as patient-derived xenograft colon and lung tumors.