Learn More
Research Products International Corp Graduated Centrifuge Tubes, 15ml, S, 50/bag, 10 bags/case
SDP
Catalog No. 502229676
Encompass
Premium quality centrifuge tubes are available in a 15ml size. High clarity polypropylene has excellent chemical and temperature resistance. Tubes withstand temperature extremes from -80°C to +122°C and can be centrifuged at maximum of 12,500 RCF(g).
Catalog No. 50-222-9676 Supplier Research Products International Corp Supplier No. 163222LC
Premium quality centrifuge tubes are available in a 15ml size. High clarity polypropylene has excellent chemical and temperature resistance. Tubes withstand temperature extremes from -80°C to +122°C and can be centrifuged at maximum of 12,500 RCF(g).