Learn More
Research Products International Corp Graduated Centrifuge Tubes, 50ml, Free Standing, Non-S, 500/cs
SDP
Catalog No. 502229682
Encompass
Premium quality centrifuge tubes are available in a 50ml size. High clarity polypropylene has excellent chemical and temperature resistance. Tubes withstand temperature extremes from -40°C to +125°C and can be centrifuged at maximum of 10,000 RCF(g).
Catalog No. 50-222-9682 Supplier Research Products International Corp Supplier No. 163265LC
Premium quality centrifuge tubes are available in a 50ml size. High clarity polypropylene has excellent chemical and temperature resistance. Tubes withstand temperature extremes from -40°C to +125°C and can be centrifuged at maximum of 10,000 RCF(g).